Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-3VVD-OH-d8 | 863971-44-2 | MFCD28386982 | 10 MG
SDP

Catalog No. 5000298984
Encompass_Preferred

Fmoc-3VVD-OH-d8 is a deuterium-labeled Fmoc-protected amino-acid derivative used as an ADC linker intermediate and as a stable-isotope tracer for analytical and pharmacokinetic studies. The deuterated analog supports mass-spectrometry quantification and may alter metabolic or pharmacokinetic behavior.

  • Deuterium-labeled analog suitable for tracer and quantification studies.
  • Used as an ADC linker intermediate in antibody-drug conjugate synthesis.
  • Formula C36H43D8N3O7; molecular weight 645.86.
  • Off-white to light yellow solid appearance.
  • Store at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month); protect from light.
  • Available in small mg-scale quantities (e.g., 10 MG).

Catalog No. 50-002-98984 Supplier Medchemexpress LLC Supplier No. HYI112910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-3VVD-OH-d8 is a deuterium-labeled Fmoc-protected amino-acid derivative used as an ADC linker intermediate and as a stable-isotope tracer for analytical and pharmacokinetic studies. The deuterated analog supports mass-spectrometry quantification and may alter metabolic or pharmacokinetic behavior.

  • Deuterium-labeled analog suitable for tracer and quantification studies.
  • Used as an ADC linker intermediate in antibody-drug conjugate synthesis.
  • Formula C36H43D8N3O7; molecular weight 645.86.
  • Off-white to light yellow solid appearance.
  • Store at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month); protect from light.
  • Available in small mg-scale quantities (e.g., 10 MG).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.