Learn More
Sigma Aldrich Fmoc-4-amino-3-hydroxybutanoic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Trichloromethanesulfenyl chloride was used in the preparation of:
- Trichloromethanesulfenyl acetate and trichloromethanesulfenyl trifluoroacetate via heterogeneous reaction with silver acetate and silver trifluoroacetate
- Trichloromethyl disulfides
- Alkyl orthocarbonates
- Trithiocarbonates
Specifications
Specifications
| Molecular Formula | C19H19NO5 |
| Linear Formula | C19H19NO5 |
| MDL Number | MFCD02682575 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 341.36 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.