Learn More
AnaSpec Fmoc-4-azidophenylalanine[Fmoc-Phe(N3)-OH | Fmoc-p-azidophenylalanine | Fmoc-phenylalanine-azide] | MW 428.44 | M.F. C24 H20 N4 O4 (1 g)
Catalog No. 50352492
Encompass
Fmoc-4-azidophenylalanine[Fmoc-Phe(N3)-OH; Fmoc-p-azidophenylalanine; Fmoc-phenylalanine-azide] ; MW 428.44; M.F. C24 H20 N4 O4 (1 g) An unusual amino acid used in Solid Phase Peptide Synthesis (SPPS) for the synthesis of photoreactive affinity labels.1,2,3 The side-chain azido (N3) group is stable in trifluoroacetic acid (TFA) or piperidine (Pip), two reagents commonly used in SPPS. The azido group can be readily converted to an amine on the solid phase or in solution via the Staudinger reduction.4
Catalog No. 50352492 Supplier AnaSpec Supplier No. AS643471000
Add to Cart
Add to Cart
Fmoc-4-azidophenylalanine[Fmoc-Phe(N3)-OH; Fmoc-p-azidophenylalanine; Fmoc-phenylalanine-azide] ; MW 428.44; M.F. C24 H20 N4 O4 (1 g) An unusual amino acid used in Solid Phase Peptide Synthesis (SPPS) for the synthesis of photoreactive affinity labels.1,2,3 The side-chain azido (N3) group is stable in trifluoroacetic acid (TFA) or piperidine (Pip), two reagents commonly used in SPPS. The azido group can be readily converted to an amine on the solid phase or in solution via the Staudinger reduction.4