Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-4-[(Methylsulfonyl)amino]-DL-Phe | 10 g | CAS 266999-20-6 | MDL MFCD01860464

Catalog No. 502352984
Encompass

Fmoc-4-[(Methylsulfonyl)amino]-DL-Phe is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 266999-20-6
- MDL: MFCD01860464
- InChIKey: XUHUSTBPCQQMFH-UHFFFAOYSA-N
- Molecular Weight: 480.535
- Molecular Formula: C25H24N2O6S
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29359090
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(methylsulfonamido)phenyl)propanoic acid
- SMILES: CS(=O)(NC1=CC=C(C=C1)CC(C(O)=O)NC(OCC2C3=CC=CC=C3C4=C2C=CC=C4)=O)=O

Catalog No. 50-235-2984 Supplier STA Pharmaceutical Supplier No. STAAA1015110GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-4-[(Methylsulfonyl)amino]-DL-Phe is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 266999-20-6
- MDL: MFCD01860464
- InChIKey: XUHUSTBPCQQMFH-UHFFFAOYSA-N
- Molecular Weight: 480.535
- Molecular Formula: C25H24N2O6S
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29359090
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(methylsulfonamido)phenyl)propanoic acid
- SMILES: CS(=O)(NC1=CC=C(C=C1)CC(C(O)=O)NC(OCC2C3=CC=CC=C3C4=C2C=CC=C4)=O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.