Learn More
Medchemexpress LLC Fmoc-7-amino-heptanoic acid | 127582-76-7 | 98.8% | 367.44 | 25 G
SDP
Catalog No. 5000452393
Encompass_Preferred
Fmoc-7-amino-heptanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups, utilized as a PROTAC linker in the synthesis of PROTACs. The Fmoc group can be deprotected under basic conditions for further conjugations, and the terminal carboxylic acid can form stable amide bonds with primary amine groups.
  • Can be utilized as a PROTAC linker
  • Fmoc group deprotects under basic conditions to yield a free amine
  • Suitable for further conjugations
  • Terminal carboxylic acid can react with primary amine groups
  • Forms stable amide bond in presence of activators (e.g., EDC or HATU)
Catalog No. 50-004-52393 Supplier Medchemexpress LLC Supplier No. 25GHYW009056
Fmoc-7-amino-heptanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups, utilized as a PROTAC linker in the synthesis of PROTACs. The Fmoc group can be deprotected under basic conditions for further conjugations, and the terminal carboxylic acid can form stable amide bonds with primary amine groups.
  • Can be utilized as a PROTAC linker
  • Fmoc group deprotects under basic conditions to yield a free amine
  • Suitable for further conjugations
  • Terminal carboxylic acid can react with primary amine groups
  • Forms stable amide bond in presence of activators (e.g., EDC or HATU)