Learn More
Medchemexpress LLC HY-W010514 100mg Medchemexpress, trans-Cyclohexane-1,2-diol CAS:1460-57-7 Purity:>98%
SDP
Catalog No. 501970675
Encompass_Preferred
Medchemexpress, HY-W007713 100mg Fmoc-8-amino-3,6-dioxaoctanoic acid CAS:166108-71-0 Fmoc-8-amino-3,6-dioxaoctanoic acid (Fmoc-NH-PEG2-CH2COOH) is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0675 Supplier Medchemexpress LLC Supplier No. HYW010514100MG
Medchemexpress, HY-W007713 100mg Fmoc-8-amino-3,6-dioxaoctanoic acid CAS:166108-71-0 Fmoc-8-amino-3,6-dioxaoctanoic acid (Fmoc-NH-PEG2-CH2COOH) is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.