Learn More
Medchemexpress LLC Fmoc-9-Aminononanoic 250mg
SDP
Catalog No. 5000205223
Encompass_Preferred
Fmoc-9-aminononanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups The compound can be used as a PROTAC linker in the synthesis of PROTACs and and other conjugation applications The Fmoc group can be deprotected under basic condition to obtain the free amine which can be used for further conjugations The terminal carboxylic acid can react with primary amine groups in the presence of activators (e g EDC or HATU) to form a stable amide bond
Catalog No. 50-002-05223 Supplier Medchemexpress LLC Supplier No. 250MGHYW111228
Fmoc-9-aminononanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups The compound can be used as a PROTAC linker in the synthesis of PROTACs and and other conjugation applications The Fmoc group can be deprotected under basic condition to obtain the free amine which can be used for further conjugations The terminal carboxylic acid can react with primary amine groups in the presence of activators (e g EDC or HATU) to form a stable amide bond