Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-9-aminononanoic acid | 50 g | CAS 212688-52-3 | MDL MFCD00273465

Catalog No. 502344429
Encompass

Fmoc-9-aminononanoic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 212688-52-3
- MDL: MFCD00273465
- InChIKey: LWISAKTUHDLUTN-UHFFFAOYSA-N
- Molecular Weight: 395.499
- Molecular Formula: C24H29NO4
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 9-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)nonanoic acid
- SMILES: O=C(NCCCCCCCCC(O)=O)OCC1C2=CC=CC=C2C3=CC=CC=C31

Catalog No. 50-234-4429 Supplier STA Pharmaceutical Supplier No. STAAA1155550GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-9-aminononanoic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 212688-52-3
- MDL: MFCD00273465
- InChIKey: LWISAKTUHDLUTN-UHFFFAOYSA-N
- Molecular Weight: 395.499
- Molecular Formula: C24H29NO4
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 9-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)nonanoic acid
- SMILES: O=C(NCCCCCCCCC(O)=O)OCC1C2=CC=CC=C2C3=CC=CC=C31

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.