Learn More
Selleck Chemical LLC Fmoc-a-d2-glycine E8241-5MG
SDP
Catalog No. 5000043018
Encompass_Preferred
Fmoc- -d2-glycine is the deuterated derivative of Fmoc-Gly-OH Fmoc-Gly-OH is a glycine derivative protected by an Fmoc group and can be used in compound synthesis
Catalog No. 50-000-43018 Supplier Selleck Chemical LLC Supplier No. E82415MG
Fmoc- -d2-glycine is the deuterated derivative of Fmoc-Gly-OH Fmoc-Gly-OH is a glycine derivative protected by an Fmoc group and can be used in compound synthesis