Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-a-Me-Lys Boc -OH, Advanced ChemTech

Catalog No. 01340303
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
01-340-303 1 g
01-340-302 5 g
2 options

Catalog No. 01-340-303

Supplier: Advanced Chem Tech FK21821G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1202003-49-3
Molecular Formula C27H34N2O6
Molecular Weight (g/mol) 482.58
MDL Number MFCD17019258
InChI Key MYQXEVZHWDILHG-MHZLTWQESA-N
IUPAC Name 6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylhexanoic acid
SMILES CC(C)(C)OC(=O)NCCCCC(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Color White to Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-a-Me-Lys Boc -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.