Learn More
Carbosynth LLC. Fmoc-Acc-Resin, 1g, Substitution: 0.3-0.7 meq/g
SDP
Catalog No. 501685712
Encompass_Preferred
Synonyms: Fmoc-7-Aminocoumarin-4-Acetic Acid-Rink Amide AM Resin, Application: Helpful Tool in Substrate Library Synthesis, Storage Temperature: 2-10°C, References: J.L. Harris, et al., PNAS, 97, 7754 (2000).
Catalog No. 50-168-5712 Supplier Carbosynth LLC. Supplier No. ACC9001PI1G
Synonyms: Fmoc-7-Aminocoumarin-4-Acetic Acid-Rink Amide AM Resin, Application: Helpful Tool in Substrate Library Synthesis, Storage Temperature: 2-10°C, References: J.L. Harris, et al., PNAS, 97, 7754 (2000).