Learn More
Medchemexpress LLC Fmoc-Ala-Ala-Asn(Trt)-OH | 1951424-92-2 | 99.8% | 738.83 g/mol | C44H42N4O7 | 1 MG
SDP
Catalog No. 5000203927
Encompass_Preferred
Fmoc-Ala-Ala-Asn(Trt)-OH is an Fmoc-protected tripeptide used as a cleavable linker precursor in the synthesis of antibody-drug conjugates (ADCs). It is supplied as a dry powder and is intended for research use in peptide/ADC assembly and linker development.
  • Used as a cleavable peptide linker precursor for ADC synthesis.
  • Fmoc-protected tripeptide suitable for solid-phase peptide synthesis or conjugation.
  • High reported purity, suitable for research applications.
  • Powder form; store under nitrogen, away from moisture.
  • Recommended storage: -80°C (2 years) or -20°C (1 year) for the powder.
Catalog No. 50-002-03927 Supplier Medchemexpress LLC Supplier No. HY1309331MG
Fmoc-Ala-Ala-Asn(Trt)-OH is an Fmoc-protected tripeptide used as a cleavable linker precursor in the synthesis of antibody-drug conjugates (ADCs). It is supplied as a dry powder and is intended for research use in peptide/ADC assembly and linker development.
  • Used as a cleavable peptide linker precursor for ADC synthesis.
  • Fmoc-protected tripeptide suitable for solid-phase peptide synthesis or conjugation.
  • High reported purity, suitable for research applications.
  • Powder form; store under nitrogen, away from moisture.
  • Recommended storage: -80°C (2 years) or -20°C (1 year) for the powder.