Learn More
Medchemexpress LLC Fmoc-Ala-Ala-PAB | 1384263-83-5 | 98.9% | 487.55 | C28H29N3O5 | 100 MG
SDP
Catalog No. 5000202099
Encompass_Preferred
Fmoc-Ala-Ala-PAB is a cleavable antibody-drug conjugate linker composed of an Fmoc-protected Ala-Ala dipeptide coupled to a para-aminobenzyl (PAB) self-immolative spacer. It is used in ADC synthesis to enable enzymatic cleavage and controlled payload release in targeted conjugates.
  • Cleavable dipeptide linker enabling enzymatic payload release.
  • Fmoc-protected N-terminus suitable for solid-phase and solution-phase synthesis.
  • Para-aminobenzyl self-immolative spacer for efficient drug release.
  • High purity suitable for conjugation chemistry.
  • Stable when stored at 4°C and protected from light.
Catalog No. 50-002-02099 Supplier Medchemexpress LLC Supplier No. HY148462100MG
Fmoc-Ala-Ala-PAB is a cleavable antibody-drug conjugate linker composed of an Fmoc-protected Ala-Ala dipeptide coupled to a para-aminobenzyl (PAB) self-immolative spacer. It is used in ADC synthesis to enable enzymatic cleavage and controlled payload release in targeted conjugates.
  • Cleavable dipeptide linker enabling enzymatic payload release.
  • Fmoc-protected N-terminus suitable for solid-phase and solution-phase synthesis.
  • Para-aminobenzyl self-immolative spacer for efficient drug release.
  • High purity suitable for conjugation chemistry.
  • Stable when stored at 4°C and protected from light.