Learn More
Carbosynth LLC. Fmoc-Ala-Gly-OH, 1g, CAS# 116747-54-7, MF= C20H20N2O5
SDP
Catalog No. 501952404
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Application: Fmoc-Protected Amino Acid for Peptide Synthesis, MW: 368.39
Catalog No. 50-195-2404 Supplier Carbosynth LLC. Supplier No. FAG1929PI1G
This item has a minimum order quantity of 2 per supplier requirements
Application: Fmoc-Protected Amino Acid for Peptide Synthesis, MW: 368.39