Learn More
Medchemexpress LLC Fmoc-Ala-OH-13C3,15N | 865720-15-6 | 99.9% | 315.30 | 5 MG
SDP
Catalog No. 5000453437
Encompass_Preferred
Fmoc-Ala-OH-13C3,15N is a 15N-labeled and 13C-labeled Fmoc-Ala-OH, which is an alanine derivative. This compound is intended for research use only and can be used as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • 15N-labeled.
  • 13C-labeled.
  • Alanine derivative.
  • For research use only.
Catalog No. 50-004-53437 Supplier Medchemexpress LLC Supplier No. HYW009204S45MG
Fmoc-Ala-OH-13C3,15N is a 15N-labeled and 13C-labeled Fmoc-Ala-OH, which is an alanine derivative. This compound is intended for research use only and can be used as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • 15N-labeled.
  • 13C-labeled.
  • Alanine derivative.
  • For research use only.