Learn More
Medchemexpress LLC Fmoc-Ala-OH-3,3,3-d3 | 225101-67-7 | 99.4% | 314.35 | 100 MG
SDP
Catalog No. 5000453150
Encompass_Preferred
Fmoc-Ala-OH-3,3,3-d3 is the deuterium labeled Fmoc-Ala-OH-3,3,3. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-53150 Supplier Medchemexpress LLC Supplier No. HYW009204S1100MG
Fmoc-Ala-OH-3,3,3-d3 is the deuterium labeled Fmoc-Ala-OH-3,3,3. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.