Learn More
Medchemexpress LLC Fmoc-Ala-Ser(psi(Me,Me)pro)-OH | 252554-78-2 | 99.7% | 438.47 | 1 G
SDP
Catalog No. 5000423891
Encompass_Preferred
Fmoc-Ala-Ser(psi(Me,Me)pro)-OH is a synthetic dipeptide compound. It is intended for research use only and is not sold to patients.
  • Dipeptide
  • Amino acid derivative
  • Stable as powder for 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-004-23891 Supplier Medchemexpress LLC Supplier No. HYP23861G
Fmoc-Ala-Ser(psi(Me,Me)pro)-OH is a synthetic dipeptide compound. It is intended for research use only and is not sold to patients.
  • Dipeptide
  • Amino acid derivative
  • Stable as powder for 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C