Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-alpha-allyl-L-alanine | 288617-71-0 | 351.40 | 1 G
SDP

Catalog No. 5000460310
Encompass_Preferred

Fmoc-alpha-allyl-L-alanine is an Fmoc-protected alanine derivative.

  • Solid appearance
  • Light yellow to yellow color
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Soluble in DMSO at ≥ 100 mg/mL (284.58 mM)

Catalog No. 50-004-60310 Supplier Medchemexpress LLC Supplier No. HYW0070521G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-alpha-allyl-L-alanine is an Fmoc-protected alanine derivative.

  • Solid appearance
  • Light yellow to yellow color
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Soluble in DMSO at ≥ 100 mg/mL (284.58 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.