Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-alpha-allyl-L-alanine | 288617-71-0 | 351.40 | 100 MG
SDP

Catalog No. 5000460397
Encompass_Preferred

Fmoc-alpha-allyl-L-alanine is an Fmoc-protected alanine derivative designed for research applications. This compound is suitable for various laboratory processes including peptide synthesis.

  • For research use only
  • Molecular weight of 351.40
  • Purity of 99.71%

Catalog No. 50-004-60397 Supplier Medchemexpress LLC Supplier No. HYW007052100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-alpha-allyl-L-alanine is an Fmoc-protected alanine derivative designed for research applications. This compound is suitable for various laboratory processes including peptide synthesis.

  • For research use only
  • Molecular weight of 351.40
  • Purity of 99.71%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.