Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-alpha-allyl-L-alanine | 288617-71-0 | 351.40 | 250 MG
SDP

Catalog No. 5000450074
Encompass_Preferred

Fmoc-alpha-allyl-L-alanine is an Fmoc-protected alanine derivative.

  • Appearance: solid
  • Color: light yellow to yellow
  • Molecular formula: C21H21NO4
  • Purity: 99.7% by HPLC
  • Enantiomeric excess: 100%
  • Optical rotation: -15.158° (C= 0.01 g/mL, DMF, 20°C, 589 nm)
  • Solid storage: 4°C, stored under nitrogen
  • In-solvent storage: -80°C for 6 months; -20°C for 1 month, stored under nitrogen

Catalog No. 50-004-50074 Supplier Medchemexpress LLC Supplier No. 250MGHYW007052
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-alpha-allyl-L-alanine is an Fmoc-protected alanine derivative.

  • Appearance: solid
  • Color: light yellow to yellow
  • Molecular formula: C21H21NO4
  • Purity: 99.7% by HPLC
  • Enantiomeric excess: 100%
  • Optical rotation: -15.158° (C= 0.01 g/mL, DMF, 20°C, 589 nm)
  • Solid storage: 4°C, stored under nitrogen
  • In-solvent storage: -80°C for 6 months; -20°C for 1 month, stored under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.