Learn More
Medchemexpress LLC Fmoc-am | 126828-35-1 | 99.8% | 539.58 | 100 G
SDP
Catalog No. 5000452378
Encompass_Preferred
FMOC-AM is a derivative of aminomethyl resin. It provides a solid-phase carrier basis for peptide chain synthesis and can be used in solid-phase peptide synthesis.
  • Derivative of aminomethyl resin
  • Provides a solid-phase carrier basis for peptide chain synthesis
  • Can be used in solid-phase peptide synthesis
Catalog No. 50-004-52378 Supplier Medchemexpress LLC Supplier No. HYW010756100G
FMOC-AM is a derivative of aminomethyl resin. It provides a solid-phase carrier basis for peptide chain synthesis and can be used in solid-phase peptide synthesis.
  • Derivative of aminomethyl resin
  • Provides a solid-phase carrier basis for peptide chain synthesis
  • Can be used in solid-phase peptide synthesis