Learn More
Medchemexpress LLC Fmoc-Am | 126828-35-1 | 99.82% | 539.58 | 50 G
SDP
Catalog No. 5000366323
Encompass_Preferred
FMOC-AM is a derivative of aminomethyl resin that provides a solid-phase carrier basis for peptide chain synthesis. It can be used in solid-phase peptide synthesis for research purposes.
  • Provides a solid-phase carrier basis for peptide chain synthesis
  • Suitable for solid-phase peptide synthesis
  • For research use only
Catalog No. 50-003-66323 Supplier Medchemexpress LLC Supplier No. HYW01075650G
FMOC-AM is a derivative of aminomethyl resin that provides a solid-phase carrier basis for peptide chain synthesis. It can be used in solid-phase peptide synthesis for research purposes.
  • Provides a solid-phase carrier basis for peptide chain synthesis
  • Suitable for solid-phase peptide synthesis
  • For research use only