Learn More
Medchemexpress LLC Fmoc-amino-PEG3-CH2COOH | 139338-72-0 | 99.8% | 429.46 | 1 G
SDP
Catalog No. 5000452687
Encompass_Preferred
Fmoc-amino-PEG3-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
Catalog No. 50-004-52687 Supplier Medchemexpress LLC Supplier No. HYW0085681G
Fmoc-amino-PEG3-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs