Learn More
Medchemexpress LLC Fmoc-amino-PEG3-CH2COOH | 139338-72-0 | 99.8% | 429.46 | 500 MG
SDP
Catalog No. 5000365821
Encompass_Preferred
Fmoc-amino-PEG3-CH2COOH is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules containing two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
Catalog No. 50-003-65821 Supplier Medchemexpress LLC Supplier No. 500MGHYW008568
Fmoc-amino-PEG3-CH2COOH is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules containing two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs