Learn More
Medchemexpress LLC Fmoc-amino-PEG5-acid | 882847-32-7 | 99.5% | 531.59 | C28H37NO9 | 1 G
SDP
Catalog No. 5000209330
Encompass_Preferred
This Fmoc-protected amino-terminated PEG5 acid is a PEG-based linker typically used in the synthesis of PROTACs and for peptide or small-molecule conjugation chemistries. It provides an Fmoc-protected amino group for orthogonal deprotection and coupling, and a five-unit PEG spacer to enhance solubility and flexibility.
  • Fmoc-protected amino group for orthogonal deprotection and amide coupling.
  • PEG5 spacer provides flexibility and improved solubility.
  • High purity demonstrated by LCMS (99.5%).
  • Molecular weight 531.59 g/mol; formula C28H37NO9.
  • Supplied as a solid; recommended storage at -20°C (long-term) or 4°C (short-term).
Catalog No. 50-002-09330 Supplier Medchemexpress LLC Supplier No. HYW0402451G
This Fmoc-protected amino-terminated PEG5 acid is a PEG-based linker typically used in the synthesis of PROTACs and for peptide or small-molecule conjugation chemistries. It provides an Fmoc-protected amino group for orthogonal deprotection and coupling, and a five-unit PEG spacer to enhance solubility and flexibility.
  • Fmoc-protected amino group for orthogonal deprotection and amide coupling.
  • PEG5 spacer provides flexibility and improved solubility.
  • High purity demonstrated by LCMS (99.5%).
  • Molecular weight 531.59 g/mol; formula C28H37NO9.
  • Supplied as a solid; recommended storage at -20°C (long-term) or 4°C (short-term).