Learn More
Medchemexpress LLC Fmoc-amino-PEG5-acid | 882847-32-7 | MFCD06656473 | 99.3% | 531.59 | C28H37NO9 | 250 MG
SDP
Catalog No. 5000209473
Encompass_Preferred
Fmoc-amino-PEG5-acid is an Fmoc-protected PEG5 linker (Fmoc-NH-PEG5-COOH) intended for use in the synthesis of PROTACs and other bifunctional conjugates. The reagent is characterized by its molecular formula (C28H37NO9), molecular weight (531.59), and CAS number (882847-32-7), and is supplied in laboratory pack sizes for synthetic applications.
  • Peg-based five-unit spacer that increases solubility and flexibility.
  • Fmoc-protected amino group enables standard Fmoc solid-phase and solution-phase coupling.
  • Terminal carboxylic acid allows amide or ester bond formation for linker attachment.
  • High reported purity supports reliable synthetic performance.
  • Available in small lab pack sizes suitable for research-scale syntheses.
Catalog No. 50-002-09473 Supplier Medchemexpress LLC Supplier No. 250MGHYW040245
Fmoc-amino-PEG5-acid is an Fmoc-protected PEG5 linker (Fmoc-NH-PEG5-COOH) intended for use in the synthesis of PROTACs and other bifunctional conjugates. The reagent is characterized by its molecular formula (C28H37NO9), molecular weight (531.59), and CAS number (882847-32-7), and is supplied in laboratory pack sizes for synthetic applications.
  • Peg-based five-unit spacer that increases solubility and flexibility.
  • Fmoc-protected amino group enables standard Fmoc solid-phase and solution-phase coupling.
  • Terminal carboxylic acid allows amide or ester bond formation for linker attachment.
  • High reported purity supports reliable synthetic performance.
  • Available in small lab pack sizes suitable for research-scale syntheses.