Learn More
Bachem Fmoc-Arg(Me)2-OH (asymmetrical)
SDP
Catalog No. 50258683
Encompass_Preferred
Fmoc-Arg(Me)2-OH (asymmetrical) Hydrochloride, 268564-10-9, C23H28N4O4, Mr 424.50, 5g. Fmoc-ADMA hydrochloride salt. Fmoc-ADMA, a building block for methylated histone sequences. The salt formation serves as temporary protection of the guanidino moiety.
Catalog No. 50-258-683 Supplier Bachem Supplier No. 4026426.0005
Fmoc-Arg(Me)2-OH (asymmetrical) Hydrochloride, 268564-10-9, C23H28N4O4, Mr 424.50, 5g. Fmoc-ADMA hydrochloride salt. Fmoc-ADMA, a building block for methylated histone sequences. The salt formation serves as temporary protection of the guanidino moiety.