Learn More
AnaSpec FMOC-ARG(ME)2-OH (ASYMMETRICAL
Supplier:  AnaSpec AS26218F1
Encompass
Fmoc-Arg(Me)2-OH (asymmetrical)[Fmoc-Nω,ω-dimethyl-L-arginine (asymmetrical)] ; MW 424.5; M.F. C23H28N4O4 (1 g)
Catalog No. 50351003