Learn More
Medchemexpress LLC HY-W013750 100mg Medchemexpress, Fmoc-Arg-OH CAS:91000-69-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW013750100MG
Encompass_Preferred
Medchemexpress, HY-W013750 100mg Fmoc-Arg-OH CAS:91000-69-0 Fmoc-Arg-OH (Fmoc-L-Arginine), an Fmoc modified Arginine, is a used in peptide synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0698