Learn More
Medchemexpress LLC Fmoc-Arg-OH | 91000-69-0 | 99.4% | 396.44 | 5 G
SDP
Catalog No. 5000454517
Encompass_Preferred
Fmoc-Arg-OH (Fmoc-L-Arginine) is an Fmoc modified Arginine used in peptide synthesis. It serves as a key raw material in the solid-phase synthesis of peptides containing arginine residues, and plays an important role in the synthesis of bradykinin and octapeptides related to the transmembrane gp41 of HIV-1.
  • Used in peptide synthesis.
  • Key raw material for solid-phase synthesis of peptides containing arginine residues.
  • Plays an important role in the synthesis of bradykinin.
  • Plays an important role in the synthesis of octapeptides related to the transmembrane gp41 of HIV-1.
Catalog No. 50-004-54517 Supplier Medchemexpress LLC Supplier No. HYW0137505G
Fmoc-Arg-OH (Fmoc-L-Arginine) is an Fmoc modified Arginine used in peptide synthesis. It serves as a key raw material in the solid-phase synthesis of peptides containing arginine residues, and plays an important role in the synthesis of bradykinin and octapeptides related to the transmembrane gp41 of HIV-1.
  • Used in peptide synthesis.
  • Key raw material for solid-phase synthesis of peptides containing arginine residues.
  • Plays an important role in the synthesis of bradykinin.
  • Plays an important role in the synthesis of octapeptides related to the transmembrane gp41 of HIV-1.