Learn More
Cayman Chemical Fmoc-Arg(Pbf)-OH, Purity: ≥98%, CAS: 154445-77-9, 10 g
SDP
Catalog No. 502912406
Encompass_Preferred
An amino acid-containing building block; has been used in solid-phase peptide synthesis
Catalog No. 50-291-2406 Supplier Cayman Chemical Supplier No. 3645810 G
An amino acid-containing building block; has been used in solid-phase peptide synthesis