Learn More
Medchemexpress LLC Fmoc-Arg(Pbf)-OH | 154445-77-9 | MFCD00235804 | 50g
SDP
Catalog No. 5000267155
Encompass_Preferred
Fmoc-Arg(Pbf)-OH is an arginine derivative containing amine protecting group Fmoc Fmoc-Arg(Pbf)-OH is a building block for the introduction of Arg into SPPS (Solid-Phase Peptide Synthesis)[1][2][3]
Catalog No. 50-002-67155 Supplier Medchemexpress LLC Supplier No. HYY163650G
Fmoc-Arg(Pbf)-OH is an arginine derivative containing amine protecting group Fmoc Fmoc-Arg(Pbf)-OH is a building block for the introduction of Arg into SPPS (Solid-Phase Peptide Synthesis)[1][2][3]