Learn More
Apexbio Technology LLC Fmoc-Arg(Pbf)-OH 154445-77-9 100g
SDP
Catalog No. 5000062329
Encompass_Preferred
Fmoc-Arg(Pbf)-OH is an arginine derivative commonly utilized in solid-phase peptide synthesis (SPPS) Its amino group is transiently protected by the 9-fluorenylmethyloxycarbonyl (Fmoc) group enabling controlled elongation of peptide chains via sequential coupling and selective Fmoc removal Concurrently its guanidine-side chain functionality is protected by the 2 2 4 6 7-pentamethyl-dihydrobenzofuran-5-sulfonyl (Pbf) protecting group designed to remain intact throughout the synthesis process but readily removable under global deprotection conditions Application fields of this derivative include peptide-based drug discovery structure-activity relationship (SAR) studies and structural biology research where precise incorporation of protected arginine is required for peptide synthesis and functional characterization
Catalog No. 50-000-62329 Supplier Apexbio Technology LLC Supplier No. A7308100000
Fmoc-Arg(Pbf)-OH is an arginine derivative commonly utilized in solid-phase peptide synthesis (SPPS) Its amino group is transiently protected by the 9-fluorenylmethyloxycarbonyl (Fmoc) group enabling controlled elongation of peptide chains via sequential coupling and selective Fmoc removal Concurrently its guanidine-side chain functionality is protected by the 2 2 4 6 7-pentamethyl-dihydrobenzofuran-5-sulfonyl (Pbf) protecting group designed to remain intact throughout the synthesis process but readily removable under global deprotection conditions Application fields of this derivative include peptide-based drug discovery structure-activity relationship (SAR) studies and structural biology research where precise incorporation of protected arginine is required for peptide synthesis and functional characterization