Learn More
Medchemexpress LLC Fmoc-Asn-Pro-Val-PABC-PNP | 2893871-64-0 | 99.3% | C43H44N6O11 | 25 MG

Supplier: Medchemexpress LLC 25MGHY144628
Fmoc-Asn-Pro-Val-PABC-PNP is a potent ADC linker, exclusively for research applications. This compound is a solid with a white to off-white appearance and has a purity of 99.3%.
- Potent ADC linker
- For research purposes only
- Appearance: solid
- Color: white to off-white
- Purity: 99.3%
- Molecular weight: 820.84
- Store at -20°C, sealed, away from moisture
- Store in solvent at -80°C for up to 6 months, or -20°C for up to 1 month
- Applicable in cancer and cancer targeted therapy research
- Classified under antibody-drug conjugate/ADC related items
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.