Learn More
Apexbio Technology LLC Fmoc-Asn(Trt)-Wang resin 25g
SDP
Catalog No. 5000062451
Encompass_Preferred
Fmoc-Asn(Trt)-Wang resin is a solid-phase resin commonly employed in Fmoc-based peptide synthesis specifically designed to facilitate the incorporation of asparagine (Asn) residues into polypeptide chains Structurally the resin comprises a Wang resin backbone anchored via the protected amino acid Fmoc-Asn(Trt) The trityl (Trt) protection group attached to the asparagine side chain stabilizes the amide functionality preventing unintended side reactions during synthetic elongation steps This resin enables precise introduction of defined peptide sequences containing asparagine residues thereby supporting studies in protein structure-function analysis bioactivity assessment disease pathogenesis investigation and pharmaceutical lead development
Catalog No. 50-000-62451 Supplier Apexbio Technology LLC Supplier No. A684525000
Fmoc-Asn(Trt)-Wang resin is a solid-phase resin commonly employed in Fmoc-based peptide synthesis specifically designed to facilitate the incorporation of asparagine (Asn) residues into polypeptide chains Structurally the resin comprises a Wang resin backbone anchored via the protected amino acid Fmoc-Asn(Trt) The trityl (Trt) protection group attached to the asparagine side chain stabilizes the amide functionality preventing unintended side reactions during synthetic elongation steps This resin enables precise introduction of defined peptide sequences containing asparagine residues thereby supporting studies in protein structure-function analysis bioactivity assessment disease pathogenesis investigation and pharmaceutical lead development