Learn More
Bachem Fmoc-Asp(2-phenylisopropyl ester)-OH
SDP
Catalog No. 50258654
Encompass_Preferred
Fmoc-Asp(2-phenylisopropyl ester)-OH, 200336-86-3, C28H27NO6, Mr 473.53, 1g. Fmoc-Asp(OPp)-OH, Fmoc-Asp(O-2-PhiPr)-OH. An aspartic acid derivative allowing specific side chain deprotection with 1% TFA in dichloromethane. Ocampo et al. applied OPp protection during the synthesis of a peptide-oligonucleotide conjugate and cleaved it with 3% trichloroacetic acid in DCM. The OPp derivative is far less prone towards base-catalyzed aspartimide formation than the OAll, OBzl, or ODmab esters, its stability comes close to the stability of the OtBu ester.
Catalog No. 50-258-654 Supplier Bachem Supplier No. 4019087.0001
Fmoc-Asp(2-phenylisopropyl ester)-OH, 200336-86-3, C28H27NO6, Mr 473.53, 1g. Fmoc-Asp(OPp)-OH, Fmoc-Asp(O-2-PhiPr)-OH. An aspartic acid derivative allowing specific side chain deprotection with 1% TFA in dichloromethane. Ocampo et al. applied OPp protection during the synthesis of a peptide-oligonucleotide conjugate and cleaved it with 3% trichloroacetic acid in DCM. The OPp derivative is far less prone towards base-catalyzed aspartimide formation than the OAll, OBzl, or ODmab esters, its stability comes close to the stability of the OtBu ester.