Learn More
Medchemexpress LLC Fmoc-Asp(CSY)-OH | 2379679-90-8 | MFCD32857296 | 99.7% | 438.50 g/mol | C23H22N2O5S | 25 MG
SDP
Catalog No. 5000282890
Encompass_Preferred
Fmoc-Asp(CSY)-OH is an Fmoc-protected aspartic acid derivative where the side-chain carboxyl is protected as a cyanosulfurylide (CSY). It is provided as a high-purity solid for use as a building block in solid-phase peptide synthesis to minimize side reactions and enable reliable incorporation of aspartic acid residues.
  • Side-chain protected as cyanosulfurylide to reduce aspartimide formation during peptide synthesis.
  • Suitable for solid-phase peptide synthesis applications.
  • High purity suitable for research use (≈99.7%).
  • Chemical formula C23H22N2O5S and molecular weight 438.50 g/mol.
  • Supplied as a solid with recommended sealed storage away from moisture at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-82890 Supplier Medchemexpress LLC Supplier No. HYW14211525MG
Fmoc-Asp(CSY)-OH is an Fmoc-protected aspartic acid derivative where the side-chain carboxyl is protected as a cyanosulfurylide (CSY). It is provided as a high-purity solid for use as a building block in solid-phase peptide synthesis to minimize side reactions and enable reliable incorporation of aspartic acid residues.
  • Side-chain protected as cyanosulfurylide to reduce aspartimide formation during peptide synthesis.
  • Suitable for solid-phase peptide synthesis applications.
  • High purity suitable for research use (≈99.7%).
  • Chemical formula C23H22N2O5S and molecular weight 438.50 g/mol.
  • Supplied as a solid with recommended sealed storage away from moisture at 4°C; in solvent: -80°C (6 months), -20°C (1 month).