Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Asp(CSY)-OH | 2379679-90-8 | MFCD32857296 | 99.7% | 438.50 g/mol | C23H22N2O5S | 50 MG
SDP

Catalog No. 5000282964
Encompass_Preferred

Fmoc-Asp(CSY)-OH is an Fmoc-protected aspartic acid derivative bearing a cyanosulfurylide (CSY) carboxyl protecting group, designed for use in peptide synthesis and amino acid derivative research. Supplied as a solid for laboratory research use only.

  • Contains a cyanosulfurylide carboxyl protecting group.
  • Compatible with Fmoc-based solid-phase peptide synthesis workflows.
  • High purity suitable for analytical and synthetic applications.
  • Solid form, white to light yellow, stable when stored sealed at 4°C.
  • Soluble in DMSO; stock solutions stable at -80°C for months or -20°C for short-term storage.
  • Available in small pack sizes suitable for building block use in peptide assembly.

Catalog No. 50-002-82964 Supplier Medchemexpress LLC Supplier No. HYW14211550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Asp(CSY)-OH is an Fmoc-protected aspartic acid derivative bearing a cyanosulfurylide (CSY) carboxyl protecting group, designed for use in peptide synthesis and amino acid derivative research. Supplied as a solid for laboratory research use only.

  • Contains a cyanosulfurylide carboxyl protecting group.
  • Compatible with Fmoc-based solid-phase peptide synthesis workflows.
  • High purity suitable for analytical and synthetic applications.
  • Solid form, white to light yellow, stable when stored sealed at 4°C.
  • Soluble in DMSO; stock solutions stable at -80°C for months or -20°C for short-term storage.
  • Available in small pack sizes suitable for building block use in peptide assembly.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.