Learn More
Bachem Fmoc-Asp(ODmab)-OH
SDP
Catalog No. 50258731
Encompass_Preferred
Fmoc-Asp(ODmab)-OH, 269066-08-2, C39H42N2O8, Mr 666.77, 1g. Dmab can be removed with 2% hydrazine in DMF generating an indazole which allows the UV-spectrophotometrical monitoring of the cleavage. Conroy et al. used this Asp derivative for the solid-phase synthesis of N-linked glycopeptides. As Dmab represents an unhindered p-substituted Z-derivative, Asp(Dmab)-containing peptides are prone to base-catalyzed aspartimide formation.
Catalog No. 50-258-731 Supplier Bachem Supplier No. 4028316.0001
Add to Cart
Add to Cart
Fmoc-Asp(ODmab)-OH, 269066-08-2, C39H42N2O8, Mr 666.77, 1g. Dmab can be removed with 2% hydrazine in DMF generating an indazole which allows the UV-spectrophotometrical monitoring of the cleavage. Conroy et al. used this Asp derivative for the solid-phase synthesis of N-linked glycopeptides. As Dmab represents an unhindered p-substituted Z-derivative, Asp(Dmab)-containing peptides are prone to base-catalyzed aspartimide formation.