Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Asp OMpe -OH, Advanced ChemTech

Catalog No. 01340544
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-544 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-340-544 Supplier Advanced Chem Tech Supplier No. FD800625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bulky protecting group designed to suppress aspartimide formation during peptide synthesis.

  • CAS Number: 180675-08-5
  • Chemical Formula: C25H29NO6
  • Application: Bulky protecting group used to suppress the side reaction of aspartimide formation.

Chemical Identifiers

CAS 180675-08-5
Molecular Formula C25H29NO6
Molecular Weight (g/mol) 439.51
InChI Key SYGKUYKLHYQKPL-UHFFFAOYNA-N
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-[(3-methylpentan-3-yl)oxy]-4-oxobutanoic acid
SMILES CCC(C)(CC)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White to Off-White
Quantity 25 g
Grade Biochemical
Chemical Name or Material Fmoc-Asp OMpe -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.