Learn More
MilliporeSigma FMOC-ASP-OTBU
Catalog No. 502301745
Encompass
Fmoc-Asp-OtBu; > /=98% by TLC and HPLC; White to light yellowish solid powder; EMD; Novabiochem; 5g; N-alpha-Fmoc-L-aspartic acid alpha-t-butyl ester; F.W. 411.5; C23H25NO6; CAS: 129460-09-9
Catalog No. 50-230-1745 Supplier MilliporeSigma Supplier No. 8520370005
Fmoc-Asp-OtBu; > /=98% by TLC and HPLC; White to light yellowish solid powder; EMD; Novabiochem; 5g; N-alpha-Fmoc-L-aspartic acid alpha-t-butyl ester; F.W. 411.5; C23H25NO6; CAS: 129460-09-9