Learn More
Bachem Fmoc-Asp(OtBu)-(Dmb)Gly-OH
SDP
Catalog No. 50312968
Encompass_Preferred
Fmoc-Asp(OtBu)-(Dmb)Gly-OH, 900152-72-9, C34H38N2O9, Mr 618.68, 1g. Dmb backbone protection completely prevents base-catalyzed aspartimide formation of the Asp(OtBu)-Gly motif during Fmoc-SPPS. The Dmb moiety is readily introduced by coupling this Fmoc-dipeptide. Contrary to the Hmb derivative, cyclic lactones cannot be formed during activation.
Catalog No. 50-312-968 Supplier Bachem Supplier No. 4062763.0001
Add to Cart
Add to Cart
Fmoc-Asp(OtBu)-(Dmb)Gly-OH, 900152-72-9, C34H38N2O9, Mr 618.68, 1g. Dmb backbone protection completely prevents base-catalyzed aspartimide formation of the Asp(OtBu)-Gly motif during Fmoc-SPPS. The Dmb moiety is readily introduced by coupling this Fmoc-dipeptide. Contrary to the Hmb derivative, cyclic lactones cannot be formed during activation.