Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Asp OtBu -OH, Advanced ChemTech

Catalog No. 01340583
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-583 100 g
01-340-579 5 g
01-340-580 25 g
01-340-581 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-340-583 Supplier Advanced Chem Tech Supplier No. FD2192100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standard Fmoc-protected derivative of aspartic acid designed for efficient peptide synthesis.

  • CAS Number: 71989-14-5
  • Molecular Weight: 411.45 g/mole
  • Formula: C23H25NO6
  • Description: Standard Fmoc-protected derivative of aspartic acid used in peptide synthesis. The t-butyl ester of the side chain is readily hydrolyzed under the same conditions used to cleave peptides from Wang resin or Rink amide resin.

Chemical Identifiers

CAS 71989-14-5
Molecular Formula C23H24NO6
Molecular Weight (g/mol) 410.45
InChI Key FODJWPHPWBKDON-IBGZPJMESA-M
IUPAC Name (2S)-4-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoate
SMILES CC(C)(C)OC(=O)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Specifications

Color White to Off-White
Quantity 100 g
Grade Biochemical
Chemical Name or Material Fmoc-Asp OtBu -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.