Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Asu(OtBu)-OH | 276869-41-1 | MFCD00080269 | ≥97.0% | 467.55 g/mol | C27H33NO6 | 250 MG
SDP

Catalog No. 5000208466
Encompass_Preferred

Fmoc-Asu(OtBu)-OH is an Fmoc-protected amino acid derivative used as a peptide synthesis intermediate. It features a tert-butyl-protected side chain carboxyl and is supplied for research use; common applications include incorporation of the asu residue during solid-phase peptide synthesis.

  • Fmoc-protected amino acid for peptide synthesis.
  • Tert-butyl protection on the side-chain carboxyl group.
  • Molecular formula C27H33NO6; molecular weight 467.55 g/mol.
  • CAS 276869-41-1 for unambiguous identification.
  • Available in multiple pack sizes, including 250 MG.
  • Provided for research use only; not for human or veterinary use.
  • Compatible with standard solid-phase peptide synthesis workflows.

Catalog No. 50-002-08466 Supplier Medchemexpress LLC Supplier No. 250MGHYW048733
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Asu(OtBu)-OH is an Fmoc-protected amino acid derivative used as a peptide synthesis intermediate. It features a tert-butyl-protected side chain carboxyl and is supplied for research use; common applications include incorporation of the asu residue during solid-phase peptide synthesis.

  • Fmoc-protected amino acid for peptide synthesis.
  • Tert-butyl protection on the side-chain carboxyl group.
  • Molecular formula C27H33NO6; molecular weight 467.55 g/mol.
  • CAS 276869-41-1 for unambiguous identification.
  • Available in multiple pack sizes, including 250 MG.
  • Provided for research use only; not for human or veterinary use.
  • Compatible with standard solid-phase peptide synthesis workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.