Learn More
Medchemexpress LLC Fmoc-azetidine-3-carboxylic acid | 193693-64-0 | MFCD01321018 | 99.9% | 323.35 | C19H17NO4 | 10 G
SDP
Catalog No. 5000204294
Encompass_Preferred
Fmoc-azetidine-3-carboxylic acid is an Fmoc-protected azetidine building block used in peptide synthesis and medicinal chemistry research. Supplied as a white to off-white solid with high chemical purity, it is suitable for incorporation of azetidine residues during solid-phase peptide synthesis or for use in linker design and small-molecule modifications. Storage recommendations and purity specifications support research and development workflows.
  • Fmoc-protected azetidine building block suitable for peptide synthesis.
  • White to off-white solid with high purity.
  • Compatible with solid-phase peptide synthesis and linker design.
  • Available in multiple pack sizes for research use.
  • Stable under recommended storage conditions for powder and solution.
Catalog No. 50-002-04294 Supplier Medchemexpress LLC Supplier No. HYW01127710G
Fmoc-azetidine-3-carboxylic acid is an Fmoc-protected azetidine building block used in peptide synthesis and medicinal chemistry research. Supplied as a white to off-white solid with high chemical purity, it is suitable for incorporation of azetidine residues during solid-phase peptide synthesis or for use in linker design and small-molecule modifications. Storage recommendations and purity specifications support research and development workflows.
  • Fmoc-protected azetidine building block suitable for peptide synthesis.
  • White to off-white solid with high purity.
  • Compatible with solid-phase peptide synthesis and linker design.
  • Available in multiple pack sizes for research use.
  • Stable under recommended storage conditions for powder and solution.