Learn More
Medchemexpress LLC Fmoc-azetidine-3-carboxylic acid | 193693-64-0 | MFCD01321018 | 99.9% | 323.35 | C19H17NO4 | 25 G
SDP
Catalog No. 5000203139
Encompass_Preferred
Fmoc-azetidine-3-carboxylic acid is an Fmoc-protected amino acid derivative used as a building block in peptide synthesis and as a linker component in ADC and PROTAC chemistry. It is supplied for research use and is characterized by defined molecular properties and high purity.
  • Fmoc-protected azetidine-3-carboxylic acid used in peptide synthesis.
  • Suitable as a cleavable ADC linker and for PROTAC linker chemistry.
  • High purity suitable for research applications.
  • Molecular formula C19H17NO4 and molecular weight 323.35.
  • Available in multiple pack sizes, including 25 g.
Catalog No. 50-002-03139 Supplier Medchemexpress LLC Supplier No. HYW01127725G
Fmoc-azetidine-3-carboxylic acid is an Fmoc-protected amino acid derivative used as a building block in peptide synthesis and as a linker component in ADC and PROTAC chemistry. It is supplied for research use and is characterized by defined molecular properties and high purity.
  • Fmoc-protected azetidine-3-carboxylic acid used in peptide synthesis.
  • Suitable as a cleavable ADC linker and for PROTAC linker chemistry.
  • High purity suitable for research applications.
  • Molecular formula C19H17NO4 and molecular weight 323.35.
  • Available in multiple pack sizes, including 25 g.