Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-Beta-Azido-Ala-Oh

Catalog No. 714151250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-415-1250MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-415-1250MG Supplier Sigma Aldrich Supplier No. 714151250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fmoc-+a-azido-Ala-OH chloride is a general N-terminal protected reagent used in the solid phase peptide synthesis. Azido group allows it to undergo copper-mediated click chemistry reactions.It can be used in:Synthesis of +a-N-acetylgalactosamine (+a-GalNAc) linked antifreeze glycopeptides (AFGPs).Synthesis of stimuli-responsive multifunctional peptide gatekeepers for drug delivery applications.Synthesis of triazole-linked glycopeptides via Cu(I)-catalyzed 1,3-dipolar cycloaddition (CuAAC).

Specifications

CAS 1293-65-8
Melting Point 52°C to 55°C
Molecular Formula C10H8Br2Fe
Linear Formula C10H8Br2Fe
MDL Number MFCD01075755
Quantity 1 g
Molecular Weight (g/mol) 343.82 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.