Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Bpa, Advanced ChemTech

Catalog No. 01340493
Click to view available options
Quantity:
1 g
5 g

Catalog No. 01-340-493

Supplier: Advanced Chem Tech FF24241G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117666-96-3
Molecular Formula C31H25NO5
Molecular Weight (g/mol) 491.54
InChI Key SYOBJKCXNRQOGA-NDEPHWFRSA-N
IUPAC Name (2S)-3-(4-benzoylphenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White/Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-Bpa
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.