Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-COP | 25 g | CAS 312965-04-1 | MDL MFCD00800355

Catalog No. 502350336
Encompass

Fmoc-COP is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 312965-04-1
- MDL: MFCD00800355
- InChIKey: TUZVFPRISNDQLQ-UHFFFAOYSA-N
- Molecular Weight: 353.374
- Molecular Formula: C20H19NO5
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 4-(((9H-fluoren-9-yl)methoxy)carbonyl)morpholine-2-carboxylic acid
- SMILES: O=C(N1CCOC(C(O)=O)C1)OCC2C3=CC=CC=C3C4=CC=CC=C42

Catalog No. 50-235-0336 Supplier STA Pharmaceutical Supplier No. STAAA1016025GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-COP is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 312965-04-1
- MDL: MFCD00800355
- InChIKey: TUZVFPRISNDQLQ-UHFFFAOYSA-N
- Molecular Weight: 353.374
- Molecular Formula: C20H19NO5
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 4-(((9H-fluoren-9-yl)methoxy)carbonyl)morpholine-2-carboxylic acid
- SMILES: O=C(N1CCOC(C(O)=O)C1)OCC2C3=CC=CC=C3C4=CC=CC=C42

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.