Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Cpa-OH | 371770-32-0 | MFCD03094886 | 99.3% | 379.45 g·mol⁻¹ | C23H25NO4 | 5 G
SDP

Catalog No. 5000209248
Encompass_Preferred

Fmoc-Cpa-OH is an Fmoc-protected β-cyclopropylalanine derivative containing both amino and carboxyl functional groups. It is supplied for research use and is commonly used as a protected building block in peptide synthesis and amino-acid modification.

  • Fmoc-protected amino acid suitable for solid-phase peptide synthesis.
  • Contains both amino and carboxyl functional groups.
  • Molecular formula C23H25NO4 and molecular weight 379.45 g·mol⁻¹.
  • High purity - 99.3% (reported).
  • CAS number 371770-32-0 for unambiguous identification.
  • Available in 1 g and 5 g packages.
  • Soluble in DMSO, as reported by the supplier.

Catalog No. 50-002-09248 Supplier Medchemexpress LLC Supplier No. HYW0419835G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Cpa-OH is an Fmoc-protected β-cyclopropylalanine derivative containing both amino and carboxyl functional groups. It is supplied for research use and is commonly used as a protected building block in peptide synthesis and amino-acid modification.

  • Fmoc-protected amino acid suitable for solid-phase peptide synthesis.
  • Contains both amino and carboxyl functional groups.
  • Molecular formula C23H25NO4 and molecular weight 379.45 g·mol⁻¹.
  • High purity - 99.3% (reported).
  • CAS number 371770-32-0 for unambiguous identification.
  • Available in 1 g and 5 g packages.
  • Soluble in DMSO, as reported by the supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.